Anti-MARK4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20174-1-AP-150UL
10089-990EA 402 USD
10089-990
Anti-MARK4 Rabbit Polyclonal Antibody
Antibodies
MARK4(MAP/microtubule affinity-regulating kinase 4) is also named as KIAA1860, MARKL1 and belongs to the CAMK Ser/Thr protein kinase family and SNF1 subfamily. It is directly involved in microtubule organization in neuronal cells and may contribute to the pathological phosphorylation of tau in Alzheimer's disease and it also plays a role in hepatocellular carcinogenesis. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 83 kDa and 75 kDa.

IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: MARK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR