Anti-KGA Rabbit Polyclonal Antibody

Supplier: Proteintech
20170-1-AP-150UL
10089-172EA 537.7 USD
10089-172
Anti-KGA Rabbit Polyclonal Antibody
Antibodies
GLS, also named as GLS1 and KIAA0838, belongs to the glutaminase family. It catalyzes the first reaction in the primary pathway for the renal catabolism of glutamine. Glutaminase-, glutamate-, and taurine-immunoreactive neurons develop neurofibrillary tangles in Alzheimer's disease.The glutaminase band in AA/C1 cells is more intense than in HT29 cells, in accordance with measurements of glutaminase activity, and had the same molecular mass of approx. 63 kDa. The bands for both cell lines are clearly different in size from both rat liver glutaminase (58 kDa) and rat kidney glutaminase (65 kDa). It also reveals a molecular weight of 83-84 kDa as a phosphate-dependent glutaminase. It has 3 isoforms produced by alternative splicing named as KGA,GAM,GAC. This antibody is specific to KGA.

IHC: Human Kidney Tissue, 1:20-1:200; Western Blot: HEK-293 Cells, 1:200-1:2000; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: KGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR