Anti-CYP4Z1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20142-1-AP-150UL
10085-544EA 402 USD
10085-544
Anti-CYP4Z1 Rabbit Polyclonal Antibody
Antibodies
CYP4Z1 belongs to the cytochrome P450 family. CYP4Z1 catalyzes the reaction: RH + reduced flavoprotein + O2 = ROH + oxidized flavoprotein + H2O. The antibody is special to CYP4Z1.

Western Blot: MDA-MB-453s Cells, 1:200-1:2000

Type: Primary
Antigen: CYP4Z1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR