Anti-CHST13 Rabbit Polyclonal Antibody

Supplier: Proteintech
20141-1-AP-150UL
10084-764EA 402 USD
10084-764
Anti-CHST13 Rabbit Polyclonal Antibody
Antibodies
CHST13, also named as C4ST3, belongs to the sulfotransferase 2 family. CHST13 catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin. Chondroitin sulfate constitutes the predominant proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. CHST13 transfers sulfate to the C4 hydroxyl of beta1,4-linked GalNAc that is substituted with a beta-linked glucuronic acid at the C-3 hydroxyl. No activity toward dermatan. The antibody is specific to CHST13.

Western Blot: Mouse Liver Tissue, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: CHST13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR