Anti-MYH4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20140-1-AP-150UL
10090-700EA 402 USD
10090-700
Anti-MYH4 Rabbit Polyclonal Antibody
Antibodies
Myosin is one of the most important skeletal muscle proteins. MYH4 functions muscle contraction. The antibody is specific to MYH4.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:1000; IF: Hela Cells, 1:10-1:100; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: MYH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR