Anti-ZHX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20136-1-AP-150UL
10097-286EA 402 USD
10097-286
Anti-ZHX2 Rabbit Polyclonal Antibody
Antibodies
  • Western Blot: Human Heart Tissue, 1:1000-1:10000

Type: Primary
Antigen: ZHX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR