Anti-ACAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20135-1-AP-150UL
10082-402EA 402 USD
10082-402
Anti-ACAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: ACAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR