Anti-C14ORF94 Rabbit Polyclonal Antibody

Supplier: Proteintech
20115-1-AP-150UL
10083-772EA 402 USD
10083-772
Anti-C14ORF94 Rabbit Polyclonal Antibody
Antibodies
HAUS4, also named as C14orf94, has four isoforms with MW 27-29 kDa, 37 kDa and 42 kDa.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: C14ORF94
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR