Anti-FLJ10357 Rabbit Polyclonal Antibody

Supplier: Proteintech
20110-1-AP-150UL
10087-060EA 402 USD
10087-060
Anti-FLJ10357 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:200-1:2000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: FLJ10357
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR