Anti-FAM96B Rabbit Polyclonal Antibody

Supplier: Proteintech
20108-1-AP-150UL
10086-844EA 402 USD
10086-844
Anti-FAM96B Rabbit Polyclonal Antibody
Antibodies
IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: FAM96B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR