Anti-EEF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20107-1-AP-150UL
10086-280EA 537.7 USD
10086-280
Anti-EEF2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Stomach cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: EEF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR