Anti-RIPK5 Rabbit Polyclonal Antibody

Supplier: Proteintech
20102-1-AP-150UL
10093-702EA 402 USD
10093-702
Anti-RIPK5 Rabbit Polyclonal Antibody
Antibodies
Protein-serine/threonine and protein-tyrosine kinases (PSK and PTK) comprise two major classes of protein kinases. They provide a key switch dictating the dynamic nature and graded regulation of signal transduction pathway activities in cellular organization. DSTYK is a members of the RIP serine/threonine kinase family which involved in activation of NF-KB, JNK, and p38, and induction of apoptosis. This antibody is specific to DSTYK.

IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: RIPK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR