Anti-CPNE9 Rabbit Polyclonal Antibody

Supplier: Proteintech
20098-1-AP-150UL
10085-130EA 402 USD
10085-130
Anti-CPNE9 Rabbit Polyclonal Antibody
Antibodies
Copines are a family of evolutionarily conserved calcium-dependent phospholipid-binding proteins . They contain two Ca(2+)- and phospholipid-binding domains known as C2 domains. Copines are potentially involved in regulating membrane trafficking and in protein-protein interactions. CPNE9, also named as KIAA4217, belongs to the copine family. This antibody is specific to CPNE9.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: CPNE9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR