Anti-CPNE8 Rabbit Polyclonal Antibody

Supplier: Proteintech
20097-1-AP-150UL
10085-128EA 402 USD
10085-128
Anti-CPNE8 Rabbit Polyclonal Antibody
Antibodies
Copines are a family of evolutionarily conserved calcium-dependent phospholipid-binding proteins . They contain two Ca(2+)- and phospholipid-binding domains known as C2 domains. Copines are potentially involved in regulating membrane trafficking and in protein-protein interactions. CPNE8 belongs to the copine family. The antibody has no cross reaction with CPNE5.

IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: CPNE8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR