Anti-LDLRAD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20096-1-AP-150UL
10089-584EA 402 USD
10089-584
Anti-LDLRAD1 Rabbit Polyclonal Antibody
Antibodies
LDLRAD1 Belongs to the LDLR family. The antibody can recognize all the 3 isoforms of LDLRAD1.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: LDLRAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR