Anti-RIM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20093-1-AP-150UL
10093-676EA 402 USD
10093-676
Anti-RIM2 Rabbit Polyclonal Antibody
Antibodies
RIMS2, also named as KIAA0751, RAB3IP3 and RIM2, is a rab effector which involved in exocytosis. RIMS2 may act as scaffold protein. The antibody is special to RIMS2. The antibody recognizes all the isoforms except isoform7 of RIMS2.

Western Blot:L02 Cells, 1:500-1:5000

Type: Primary
Antigen: RIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR