Anti-RIMS4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20091-1-AP-150UL
10093-682EA 402 USD
10093-682
Anti-RIMS4 Rabbit Polyclonal Antibody
Antibodies
RIMS4, also named as C20orf190, regulates synaptic membrane exocytosis. The antibody is specific to RIMS4.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: RIMS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR