Anti-DGKK Rabbit Polyclonal Antibody

Supplier: Proteintech
20090-1-AP-150UL
10085-798EA 402 USD
10085-798
Anti-DGKK Rabbit Polyclonal Antibody
Antibodies
DGKK Belongs to the eukaryotic diacylglycerol kinase family. DGKK phosphorylates diacylglycerol (DAG) to generate phosphatidic acid (PA). DGKK catalyze the reaction: ATP + 1,2-diacylglycerol = ADP + 1,2-diacyl-sn-glycerol 3-phosphate. The antibody is specific to DGKK.

Western Blot: Human Testis Tissue, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: DGKK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR