Anti-GAB3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20089-1-AP-150UL
10087-220EA 402 USD
10087-220
Anti-GAB3 Rabbit Polyclonal Antibody
Antibodies
GAB3 belongs to the GAB family. GAB3 interacts with PIK3R/p85, SHP2 and GRAP2/MONA. It may interact with Grb2. The antibody is specific to GAB3.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GAB3-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR