Anti-MAS1L Rabbit Polyclonal Antibody

Supplier: Proteintech
20080-1-AP-150UL
10089-644EA 402 USD
10089-644
Anti-MAS1L Rabbit Polyclonal Antibody
Antibodies
MAS1L, also named as MRG, belongs to the G-protein coupled receptor 1 family and Mas subfamily. The antibody is specific to MAS1L.

Western Blot: HEK-293 Cells, 1:1000-1:10000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: MAS1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR