Anti-MYSM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20078-1-AP-150UL
10090-782EA 402 USD
10090-782
Anti-MYSM1 Rabbit Polyclonal Antibody
Antibodies
MYSM1, also named as KIAA1915, belongs to the peptidase M67A family and MYSM1 subfamily. MYSM1 is a metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. MYSM1 acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation. It catalyzes the reaction: Ubiquitin C-terminal thioester + H2O = ubiquitin + a thiol. MYSM1 is a marker of cancer. The antibody is specific to MYSM1.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human spleen, 1:20-1:200

Type: Primary
Antigen: MYSM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR