Anti-GPR101 Rabbit Polyclonal Antibody

Supplier: Proteintech
20074-1-AP-150UL
10087-764EA 402 USD
10087-764
Anti-GPR101 Rabbit Polyclonal Antibody
Antibodies
GPR101, also named as GPCR6, belongs to the G-protein coupled receptor 1 family. It is an orphan receptor. The antibody is special to GPR101.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: GPR101-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR