Anti-BAGE4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20071-1-AP-150UL
10083-492EA 402 USD
10083-492
Anti-BAGE4 Rabbit Polyclonal Antibody
Antibodies
BAGE4, also named as MLL3P and CT2.4, belongs to the BAGE family. It is a candidate gene encoding tumor antigens. BAGE4 is Cancer/testis antigen 2.4. The antibody can recognize BAGE4, BAGE, BAGE3, BAGE5.

IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: BAGE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR