Anti-PCDH11Y Rabbit Polyclonal Antibody

Supplier: Proteintech
20069-1-AP-150UL
10091-918EA 402 USD
10091-918
Anti-PCDH11Y Rabbit Polyclonal Antibody
Antibodies
PCDH11Y, also named as PCDH11, PCDH22 and PCDHY, is a potential calcium-dependent cell-adhesion protein. The antibody can recognize PCDH11X and PCDH11Y.

Western Blot: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: PCDH11Y
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR