Anti-NOX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20065-1-AP-150UL
10091-230EA 402 USD
10091-230
Anti-NOX3 Rabbit Polyclonal Antibody
Antibodies
NOX3 is a member of NADPH oxidase which constitutively produces superoxide upon formation of a complex with CYBA/p22phox. NOX3 plays a role in the biogenesis of otoconia/otolith, which are crystalline structures of the inner ear involved in the perception of gravity. The antibody is specific to NOX3.

Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: NOX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR