Anti-TRERF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20062-1-AP-150UL
10096-306EA 402 USD
10096-306
Anti-TRERF1 Rabbit Polyclonal Antibody
Antibodies
TRERF1, also named as BCAR2, RAPA and TREP132, activates transcription of CYP11A1. TRERF1 interaction with CREBBP and EP300 results in a synergistic transcriptional activation of CYP11A1. The antibody recognizes the C-term of TRERF1.

Western Blot: MCF7 Cells, 1:200-1:1000

Type: Primary
Antigen: TRERF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR