Anti-TNS3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20053-1-AP-150UL
10096-166EA 402 USD
10096-166
Anti-TNS3 Rabbit Polyclonal Antibody
Antibodies
TNS3, also named as TEM6, TENS1 and TPP, may play a role in actin remodeling. It is involved in the dissociation of the integrin-tensin-actin complex. EGF activates TNS4 and down-regulates TNS3 which results in capping the tail of ITGB1. TNS3 seems to be involved in mammary cell migration. It may be involved in cell migration and bone development. The antibody recognizes all the isoforms of TNS3.

Western Blot: Mouse Kidney Tissue, 1:200-1:2000

Type: Primary
Antigen: TNS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR