Anti-MPC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20049-1-AP-150UL
10090-364EA 537.7 USD
10090-364
Anti-MPC2 Rabbit Polyclonal Antibody
Antibodies
BRP44 belongs to the UPF0041 (0-44) family. The antibody is specific to BRP44.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR