Anti-DAPK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20048-1-AP-150UL
10085-660EA 402 USD
10085-660
Anti-DAPK2 Rabbit Polyclonal Antibody
Antibodies
DAPK2, also named as DRP-1, belongs to the protein kinase superfamily, CAMK Ser/Thr protein kinase family and DAP kinase subfamily. DAPK2 is a calcium/calmodulin-dependent serine/threonine kinase which acts as a positive regulator of apoptosis. DAPK2 catalyzes the reaction: ATP + a protein = ADP + a phosphoprotein. The antibody is specific to DAPK2.

Western Blot:A431 Cells, 1:200-1:1000; IF:A375 Cells, 1:10-1:100

Type: Primary
Antigen: DAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR