Anti-LPHN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20045-1-AP-150UL
10089-772EA 402 USD
10089-772
Anti-LPHN3 Rabbit Polyclonal Antibody
Antibodies
LPHN3, also named as KIAA0768 and LEC3, belongs to the G-protein coupled receptor 2 family and LN-TM7 subfamily. LPHN3 forms a heterodimer, consisting of a large extracellular region (p120) non-covalently linked to a seven-transmembrane moiety (p85). The antibody recognizes all the isoforms of LPHN3. the antibody has no cross reaction to LPHN1 and LPHN2.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: LPHN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR