Anti-ARHGEF9 Rabbit Polyclonal Antibody

Supplier: Proteintech
20042-1-AP-150UL
10083-106EA 402 USD
10083-106
Anti-ARHGEF9 Rabbit Polyclonal Antibody
Antibodies
ARHGEF9, also named as KIAA0424, acts as guanine nucleotide exchange factor (GEF) for CDC42. ARHGEF9 promotes formation of GPHN clusters. The antibody is special to ARHGEF9.

Western Blot: Human Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: ARHGEF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR