Anti-ZC3H4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20041-1-AP-150UL
10097-252EA 524.48 USD
10097-252
Anti-ZC3H4 Rabbit Polyclonal Antibody
Antibodies
ZC3H4, also named as C19orf7 and KIAA1064, is a 140kd protein. The antibody can recognizes near the C-term of ZC3H4.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: ZC3H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR