Anti-MPRIP Rabbit Polyclonal Antibody

Supplier: Proteintech
20040-1-AP-150UL
10090-392EA 402 USD
10090-392
Anti-MPRIP Rabbit Polyclonal Antibody
Antibodies
MPRIP, also named as KIAA0864, MRIP, RHOIP3 and p116Rip, is required for the regulation of the actin cytoskeleton by RhoA and ROCK1. MRPIP targets myosin phosphatase to the actin cytoskeleton. Depletion leads to an increased number of stress fibers in smooth muscle cells through stabilization of actin fibers by phosphorylated myosin. Overexpression of MRIP as well as its F-actin-binding region leads to disassembly of stress fibers in neuronal cells. The antibody recognizes the C-term of isoform2 and isoform3 of MPRIP.

Western Blot:SH-SY5Y Cells, 1:200-1:2000; IHC: Human ovary Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: MPRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR