Anti-PDZD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20039-1-AP-150UL
10092-064EA 402 USD
10092-064
Anti-PDZD2 Rabbit Polyclonal Antibody
Antibodies
PDZD2, also named as AIPC, KIAA0300 and PDZK3, may be associated with the early promotion of prostate tumoregenesis. The antibody recognizes near the N-term of PDZD2.

IHC: Human lung, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: PDZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR