Anti-ANKRD26 Rabbit Polyclonal Antibody

Supplier: Proteintech
20035-1-AP-150UL
10082-906EA 402 USD
10082-906
Anti-ANKRD26 Rabbit Polyclonal Antibody
Antibodies
ANKRD26 also named as KIAA1074. The antibody recognizes the C-term of ANKRD26.

IHC: Human breast cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: ANKRD26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR