Anti-CIT Rabbit Polyclonal Antibody

Supplier: Proteintech
20033-1-AP-150UL
10084-606EA 402 USD
10084-606
Anti-CIT Rabbit Polyclonal Antibody
Antibodies
CIT, also named KIAA0949, STK21 and CRIK, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. It is required for KIF14 localization to the central spindle and midbody. CIT may play a role in cytokinesis. Putative RHO/RAC effector that binds to the GTP-bound forms of RHO and RAC1. It probably binds p21 with a tighter specificity in vivo. CIT plays an important role in the regulation of cytokinesis and the development of the central nervous system. The antibody recognizes all the isoformes of CIT.

Western Blot:A431 Cells, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR