Anti-LAMC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20030-1-AP-150UL
10089-484EA 402 USD
10089-484
Anti-LAMC3 Rabbit Polyclonal Antibody
Antibodies
Binding to cells via a high affinity receptor, LAMC3 is thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. The antibody is specific to LAMC3.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: LAMC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR