Anti-DLEC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20027-1-AP-150UL
10085-888EA 402 USD
10085-888
Anti-DLEC1 Rabbit Polyclonal Antibody
Antibodies
DLEC1, also named as DLC1 F56, may act as a tumor suppressor by inhibiting cell proliferation.

Western Blot: Human Testis Tissue, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: DLEC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR