Anti-ZBTB40 Rabbit Polyclonal Antibody

Supplier: Proteintech
20026-1-AP-150UL
10097-248EA 402 USD
10097-248
Anti-ZBTB40 Rabbit Polyclonal Antibody
Antibodies
ZBTB40, also named as KIAA0478, belongs to the krueppel C2H2-type zinc-finger protein family. ZBTB40 may be involved in transcriptional regulation. The antibody can recognizes all the isoforms of ZBTB40.

Western Blot:A431 Cells, 1:500-1:5000

Type: Primary
Antigen: ZBTB40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR