Anti-URB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20023-1-AP-150UL
10096-804EA 402 USD
10096-804
Anti-URB1 Rabbit Polyclonal Antibody
Antibodies
URB1, also named as C21orf108, KIAA0539 and NPA1, is a 2,271 amino acid protein that is homologous to the S. cerevisiae protein URB1 which is a nucleolar protein involved in the normal accumulation of 25S and 5.8S rRNAs and may also play a role in the biosynthesis of the 60S ribosomal unit. Also localized to the nucleolus, human URB1 is thought to have the same functions as S. cerevisiae URB1. The antibody is specific to URB1.

Western Blot:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: URB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR