Anti-GPRASP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20020-1-AP-150UL
10087-800EA 402 USD
10087-800
Anti-GPRASP1 Rabbit Polyclonal Antibody
Antibodies
GPRASP1, also named as GASP, GASP1 and KIAA0443, belongs to the GPRASP family. GPRASP1 may modulate lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. It is targets receptors for degradation in lysosomes. The antibody is specific to GPRASP1.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: GPRASP1-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR