Anti-CYP4F11 Rabbit Polyclonal Antibody

Supplier: Proteintech
20012-1-AP-150UL
10081-208EA 402 USD
10081-208
Anti-CYP4F11 Rabbit Polyclonal Antibody
Antibodies
CYP4F11 belongs to the cytochrome P450 family. Cytochromes P450 are a group of heme-thiolate monooxygenases. CYP4F11 catalyze the reaction: RH + reduced flavoprotein + O2 = ROH + oxidized flavoprotein + H2O. The antibody is specific to CYP4F11. It has no cross reaction to other CYP4F member.This antibody is specific to CYP4F11.

Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CYP4F11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR