Anti-CD134/OX40 Rabbit Polyclonal Antibody

Supplier: Proteintech
20006-1-AP-150UL
10083-330EA 402 USD
10083-330
Anti-CD134/OX40 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: CD134/OX40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR