Anti-TGFBR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20000-1-AP-150UL
10095-862EA 402 USD
10095-862
Anti-TGFBR3 Rabbit Polyclonal Antibody
Antibodies
TGFBR3, also named as TGFR-3, BGCAN and betaglycan, binds to TGF-beta. TGFBR3 could be involved in capturing and retaining TGF-beta for presentation to the signaling receptors. The antibody is special to TGFBR3.

Western Blot:A549 Cells, 1:500-1:5000

Type: Primary
Antigen: TGFBR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR