Anti-TGF-beta 2-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
19999-1-AP-150UL
10095-854EA 537.7 USD
10095-854
Anti-TGF-beta 2-Specific Rabbit Polyclonal Antibody
Antibodies
Tranforming growth factor beta-2 (TGFB2), also named G-TSF, BSC-1 cell growth inhibitor, Polyergin and Cetermin, belongs to the TGF-beta family. TGFB2 is a secreted protein and contains a 19aa signal peptides at its N-terminal. Heterodimers with TGFB1 and with TGFB2 have been found in bone. It was proved to have suppressive effects on interleukin-2 dependent T-cell growth. This antibody is TGFB2 specific not detecting other TGFBs.

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: TGF-beta 2-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR