Anti-HBQ1 Rabbit Polyclonal Antibody

Supplier: Proteintech
19997-1-AP-150UL
10088-084EA 402 USD
10088-084
Anti-HBQ1 Rabbit Polyclonal Antibody
Antibodies
HBQ1, also named as Theta-1-globin, belongs to the globin family. The antibody is specific to HBQ1.

Western Blot:recombinant Protein, 1:1000-1:10000

Type: Primary
Antigen: HBQ1-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR