Anti-HBZ Rabbit Polyclonal Antibody

Supplier: Proteintech
19995-1-AP-150UL
10088-092EA 402 USD
10088-092
Anti-HBZ Rabbit Polyclonal Antibody
Antibodies
HBZ, also named as Zeta-globin and HBAZ, belongs to the globin family. HBZ is an alpha-type chain of mammalian embryonic hemoglobin, synthesized primarily in the yolk sac. The antibody is specific to HBZ.

Western Blot:recombinant Protein, 1:1000-1:10000

Type: Primary
Antigen: HBZ-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR