Anti-LDHC Rabbit Polyclonal Antibody

Supplier: Proteintech
19989-1-AP-150UL
10089-578EA 402 USD
10089-578
Anti-LDHC Rabbit Polyclonal Antibody
Antibodies
LDHC, also named as CT32 and LDH-X, belongs to the LDH/MDH superfamily and LDH family. LDHC is a possible role in sperm motility. LDHC catalyzes the reaction: (S)-lactate + NAD+ = pyruvate + NADH. LDHC is a testis-specific isoform. Abnormal LDHC expression is associated with several forms of cancer. The antibody has no cross reaction with LDHA and LDHB.

IHC: Human Testis Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; Western Blot: Human Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: LDHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR