Anti-LDHA Rabbit Polyclonal Antibody

Supplier: Proteintech
19987-1-AP-150UL
10089-572EA 402 USD
10089-572
Anti-LDHA Rabbit Polyclonal Antibody
Antibodies
LDHA, also named as PIG19, belongs to the LDH/MDH superfamily and LDH family. Defects in LDHA are the cause of glycogen storage disease type 11 (GSD11).It has 5 isoforms and can form a homotetramer .The antibody has no cross reaction to LDHB and LDHC.This antibody is specific to LDHA.

Western Blot:A431 Cells, 1:1000-1:10000; IHC: Human Heart Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR