Anti-NKTR Rabbit Polyclonal Antibody

Supplier: Proteintech
19978-1-AP-150UL
10091-132EA 402 USD
10091-132
Anti-NKTR Rabbit Polyclonal Antibody
Antibodies
NKTR is a component of a putative tumor-recognition complex. It is involved in the function of NK cells.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human spleen, 1:20-1:200

Type: Primary
Antigen: NKTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR